siRNA Details
Gene Symbol: AKAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004842.3 | NP_004833.1 | 2 | 943 | ||
NM_016377.3 | NP_057461.2 | 8 | 1413 | |||
NM_138633.2 | NP_619539.1 | 3 | 1012 | |||
GenBank mRNA | AF152929.1 | - | 7 | 1280 | ||
BC016927.1 | - | 2 | 621 | |||
BC063499.1 | - | 2 | 677 | |||
BC073847.1 | - | 2 | 679 | |||
BC094732.1 | - | 2 | 681 | |||
BC110897.1 | - | 2 | 635 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
