siRNA Details
Gene Symbol: AKAP7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004842.3 | NP_004833.1 | 2 | 2314 | ||
NM_016377.3 | NP_057461.2 | 8 | 2784 | |||
NM_138633.2 | NP_619539.1 | 3 | 2383 | |||
XM_005267228.1 | XP_005267285.1 | 8 | 1870 | |||
XM_005267229.4 | XP_005267286.1 | 8 | 2790 | |||
XM_017011508.1 | XP_016866997.1 | 8 | 2412 | |||
XM_017011509.1 | XP_016866998.1 | 8 | 1918 | |||
XM_017011510.1 | XP_016866999.1 | 7 | 1728 | |||
XM_017011511.1 | XP_016867000.1 | 6 | 1653 | |||
GenBank mRNA | AF152929.1 | - | 7 | 2651 | ||
BC016927.1 | - | 2 | 1992 | |||
BC063499.1 | - | 2 | 2048 | |||
BC094732.1 | - | 2 | 2052 | |||
BC110897.1 | - | 2 | 2006 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
