siRNA Details
Gene Symbol: AP2S1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301076.1 | NP_001288005.1 | 4 | 468 | ||
NM_001301078.1 | NP_001288007.1 | 4 | 585 | |||
NM_001301081.1 | NP_001288010.1 | 4 | 425 | |||
NM_004069.4 | NP_004060.2 | 4 | 543 | |||
NM_021575.3 | NP_067586.1 | 3 | 429 | |||
XM_011526423.2 | XP_011524725.1 | 4 | 521 | |||
XM_011526424.2 | XP_011524726.1 | 4 | 416 | |||
GenBank mRNA | AJ010148.1 | - | 3 | 314 | ||
AJ010149.1 | - | 2 | 200 | |||
AJ713543.1 | - | 3 | 236 | |||
AK312003.1 | - | 4 | 395 | |||
BC006337.2 | - | 4 | 352 | 4304127 | ||
BF970609.1 | - | 146 | ||||
BG282225.1 | - | 4 | 393 | |||
BI561502.1 | - | 4 | 430 | |||
BQ221562.1 | - | 4 | 468 | |||
BQ684675.1 | - | 1 | 18 | |||
CA454481.1 | - | 4 | 458 | |||
X97074.1 | - | 4 | 428 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
