siRNA Details
Gene Symbol: DNAJC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256864.1 | NP_001243793.1 | 19 | 5656 | ||
NM_001256865.1 | NP_001243794.1 | 20 | 5604 | |||
NM_014787.3 | NP_055602.1 | 19 | 5491 | |||
GenBank mRNA | AB007942.1 | - | 19 | 5435 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
