siRNA Details
Gene Symbol: CEP126 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020802.3 | NP_065853.3 | 7 | 3130 | ||
XM_005271627.4 | XP_005271684.2 | 7 | 2861 | |||
GenBank mRNA | AB037798.1 | - | 4 | 2473 | ||
AK095004.1 | - | 6 | 2760 | |||
AK296350.1 | - | 4 | 2871 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
