siRNA Details
Gene Symbol: MTIF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005369.1 | NP_001005369.1 | 6 | 1008 | ||
NM_001321001.1 | NP_001307930.1 | 5 | 543 | |||
NM_001321002.1 | NP_001307931.1 | 5 | 515 | |||
NM_001321003.1 | NP_001307932.1 | 6 | 725 | |||
NM_001321004.1 | NP_001307933.1 | 4 | 777 | |||
NM_001321005.1 | NP_001307934.1 | 5 | 587 | |||
NM_002453.2 | NP_002444.2 | 5 | 587 | |||
XM_005264335.3 | XP_005264392.1 | 3 | 386 | |||
XM_011532871.2 | XP_011531173.1 | 4 | 420 | |||
XM_017004162.1 | XP_016859651.1 | 6 | 716 | |||
XM_017004163.1 | XP_016859652.1 | 5 | 792 | |||
XM_017004164.1 | XP_016859653.1 | 5 | 581 | |||
XM_017004165.1 | XP_016859654.1 | 6 | 780 | |||
XM_017004166.1 | XP_016859655.1 | 6 | 972 | |||
XM_017004167.1 | XP_016859656.1 | 1 | 32 | |||
GenBank mRNA | AF494407.1 | - | 5 | 567 | ||
AK225184.1 | - | 5 | 556 | |||
AK307993.1 | - | 5 | 567 | |||
BC039848.1 | - | 5 | 545 | |||
BC064977.1 | - | 5 | 527 | |||
BP282053.1 | - | 5 | 495 | |||
CB961193.1 | - | 5 | 610 | |||
L34600.1 | - | 5 | 541 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
