siRNA Details
Gene Symbol: SPCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014752.2 | NP_055567.2 | 5 | 1336 | ||
GenBank mRNA | AK293128.1 | - | 5 | 1317 | ||
BC008063.2 | - | 1 | 1197 | 2967488 | ||
BC070276.1 | - | 1 | 1074 | 4656138 | ||
BC106066.1 | - | 5 | 1300 | 4521501 | ||
D14658.1 | - | 5 | 1295 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
