siRNA Details
Gene Symbol: SPCS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014752.2 | NP_055567.2 | 1, 2 | 145 | ||
GenBank mRNA | AK293128.1 | - | 1, 2 | 126 | ||
BC064957.1 | - | 1, 2 | 110 | 6092500 | ||
BC106066.1 | - | 1, 2 | 110 | 4521501 | ||
BP338843.1 | - | 1, 2 | 145 | |||
D14658.1 | - | 1, 2 | 104 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
