siRNA Details
Gene Symbol: NUP160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318399.1 | NP_001305328.1 | 3 | 972 | ||
NM_015231.2 | NP_056046.1 | 3, 4 | 660 | |||
NR_134636.1 | 3, 4 | 660 | ||||
GenBank mRNA | AK026236.1 | - | 3 | 755 | ||
AK304308.1 | - | 3, 4 | 446 | |||
AL832029.1 | - | 3, 4 | 450 | |||
BC009822.2 | - | 3 | 933 | |||
BC125227.1 | - | 3, 4 | 583 | 40124592 | ||
BC125228.1 | - | 3, 4 | 583 | 40124593 | ||
BX647634.1 | - | 3, 4 | 445 | |||
D83781.1 | - | 3, 4 | 429 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
