siRNA Details
Gene Symbol: NUP160 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015231.2 | NP_056046.1 | 19 | 2536 | ||
NR_134636.1 | 19 | 2551 | ||||
GenBank mRNA | AK304308.1 | - | 19 | 2322 | ||
AL832029.1 | - | 19 | 2326 | |||
BC125227.1 | - | 19 | 2459 | 40124592 | ||
BC125228.1 | - | 19 | 2459 | 40124593 | ||
BX647634.1 | - | 19 | 2336 | |||
D83781.1 | - | 19 | 2305 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
