siRNA Details
Gene Symbol: GREB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014668.3 | NP_055483.2 | 5 | 900 | ||
NM_033090.2 | NP_149081.1 | 5 | 851 | |||
NM_148903.2 | NP_683701.2 | 5 | 916 | |||
XM_005246192.4 | XP_005246249.1 | 5 | 922 | |||
XM_011510418.2 | XP_011508720.1 | 4 | 761 | |||
XM_011510419.2 | XP_011508721.1 | 4 | 761 | |||
XM_011510423.2 | XP_011508725.1 | 4 | 761 | |||
GenBank mRNA | AF245388.1 | - | 5 | 900 | ||
AF245389.1 | - | 5 | 851 | |||
AF245390.1 | - | 5 | 916 | |||
BC071853.1 | - | 4 | 581 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
