siRNA Details
Gene Symbol: RNF103-CHMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198954.1 | NP_001185883.1 | 8 | 1075 | ||
GenBank mRNA | AK294389.1 | - | 8 | 1075 |
Gene Symbol: CHMP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005753.2 | NP_001005753.1 | 5 | 833 | ||
NM_001193517.1 | NP_001180446.1 | 6 | 774 | |||
NM_016079.3 | NP_057163.1 | 6 | 894 | |||
NR_036454.1 | 4 | 653 | ||||
GenBank mRNA | AF151907.1 | - | 6 | 763 | ||
AK025260.1 | - | 6 | 792 | |||
AK129502.1 | - | 6 | 651 | |||
AK290725.1 | - | 5 | 724 | |||
AK315835.1 | - | 5 | 777 | |||
AY364249.1 | - | 6 | 745 | |||
BC004419.1 | - | 6 | 792 | 3660116 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
