siRNA Details
Gene Symbol: PKP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000299.3 | NP_000290.2 | 8 | 1585 | ||
NM_001005337.2 | NP_001005337.1 | 7 | 1522 | |||
GenBank mRNA | AK315291.1 | - | 7 | 1343 | ||
BC114571.1 | - | 7 | 1405 | 40082609 | ||
BC115702.1 | - | 7 | 1167 | |||
X79293.1 | - | 7 | 1459 | |||
Z34974.1 | - | 7 | 1523 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
