siRNA Details
Gene Symbol: RAB3GAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172435.1 | NP_001165906.1 | 2 | 76 | ||
NM_012233.2 | NP_036365.1 | 2 | 76 | |||
XM_011510823.2 | XP_011509125.1 | 2 | 76 | |||
XM_011510825.2 | XP_011509127.1 | 2 | 76 | |||
GenBank mRNA | AB449877.1 | - | 2 | 33 | ||
AK022494.1 | - | 2 | 52 | |||
BC146809.1 | - | 2 | 46 | 8860368 | ||
BP872264.1 | - | 2 | 57 | |||
D31886.1 | - | 2 | 35 | |||
DA333844.1 | - | 2 | 76 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
