siRNA Details
Gene Symbol: ACTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103.3 | NP_001094.1 | 21 | 3637 | ||
NM_001278343.1 | NP_001265272.1 | 21 | 3637 | |||
NM_001278344.1 | NP_001265273.1 | 23 | 3834 | |||
GenBank mRNA | M86406.1 | - | 21 | 3592 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
