siRNA Details
Gene Symbol: ACTN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001103.3 | NP_001094.1 | 21 | 2920 | ||
NM_001278343.1 | NP_001265272.1 | 21 | 2920 | |||
NM_001278344.1 | NP_001265273.1 | 23 | 3117 | |||
GenBank mRNA | AB209521.1 | - | 23 | 3061 | ||
AK294906.1 | - | 19 | 2431 | |||
AK297469.1 | - | 15 | 2065 | |||
AK297618.1 | - | 14 | 2002 | |||
AK297628.1 | - | 14 | 1780 | |||
BC047901.2 | - | 21 | 2852 | 6199155 | ||
BC051770.1 | - | 21 | 2865 | 6198688 | ||
M86406.1 | - | 21 | 2873 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
