siRNA Details
Gene Symbol: ARHGAP31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020754.3 | NP_065805.2 | 7, 8 | 1407 | ||
XM_006713714.3 | XP_006713777.1 | 7, 8 | 1491 | |||
XM_017006955.1 | XP_016862444.1 | 6, 7 | 1346 | |||
GenBank mRNA | AB033030.1 | - | 7, 8 | 1407 | ||
AK293726.1 | - | 7, 8 | 1004 | |||
BC112163.1 | - | 7, 8 | 902 | 8327631 | ||
BC112165.1 | - | 7, 8 | 902 | 8327633 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
