siRNA Details
Gene Symbol: SUMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005782.1 | NP_001005782.1 | 4 | 465 | ||
NM_003352.4 | NP_003343.1 | 5 | 540 | |||
GenBank mRNA | AB062294.1 | - | 5 | 516 | ||
BC005899.1 | - | 5 | 508 | |||
BC006462.1 | - | 5 | 524 | 3546125 | ||
BC053528.1 | - | 5 | 498 | 3452929 | ||
BC066306.1 | - | 5 | 516 | 4818792 | ||
BG569821.1 | - | 4 | 434 | |||
U38784.1 | - | 6 | 458 | |||
U61397.1 | - | 5 | 537 | |||
U67122.1 | - | 5 | 472 | |||
U83117.1 | - | 5 | 527 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
