siRNA Details
Gene Symbol: HERC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271602.1 | NP_001258531.1 | 25 | 3739 | ||
NM_014606.2 | NP_055421.1 | 26 | 3994 | |||
XM_005263327.3 | XP_005263384.1 | 27 | 4121 | |||
XM_005263329.3 | XP_005263386.1 | 25 | 3970 | |||
XM_005263330.3 | XP_005263387.1 | 25 | 3823 | |||
XM_011532397.2 | XP_011530699.1 | 13 | 2234 | |||
XM_017008806.1 | XP_016864295.1 | 26 | 4097 | |||
XM_017008807.1 | XP_016864296.1 | 24 | 3693 | |||
XM_017008808.1 | XP_016864297.1 | 19 | 2896 | |||
GenBank mRNA | BQ925135.1 | - | 1 | 598 | ||
D25215.1 | - | 26 | 3921 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
