siRNA Details
Gene Symbol: HERC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271602.1 | NP_001258531.1 | 7 | 811 | ||
NM_001318505.1 | NP_001305434.1 | 8 | 1066 | |||
NM_014606.2 | NP_055421.1 | 8 | 1066 | |||
XM_005263327.3 | XP_005263384.1 | 9 | 1193 | |||
XM_005263329.3 | XP_005263386.1 | 8 | 1066 | |||
XM_005263330.3 | XP_005263387.1 | 8 | 1066 | |||
XM_005263331.1 | XP_005263388.1 | 8 | 1066 | |||
XM_005263332.3 | XP_005263389.1 | 8 | 1066 | |||
XM_017008806.1 | XP_016864295.1 | 9 | 1193 | |||
XM_017008807.1 | XP_016864296.1 | 7 | 789 | |||
GenBank mRNA | AK289990.1 | - | 8 | 1009 | ||
AK296379.1 | - | 7 | 787 | |||
BC038960.1 | - | 8 | 1004 | |||
D25215.1 | - | 8 | 993 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
