siRNA Details
Gene Symbol: PCDH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032961.2 | NP_116586.1 | 3 | 3585 | ||
XM_011532150.1 | XP_011530452.1 | 3 | 3585 | |||
GenBank mRNA | AB037821.1 | - | 3 | 3560 | ||
AL157470.1 | - | 3 | 642 | |||
BC111560.1 | - | 3 | 2923 | 40068151 | ||
KJ534913.1 | - | 3 | 2734 | |||
KJ534914.1 | - | 4 | 2563 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
