siRNA Details
Gene Symbol: PCDH10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020815.2 | NP_065866.1 | 1 | 1804 | ||
NM_032961.2 | NP_116586.1 | 1 | 1804 | |||
XM_011532150.1 | XP_011530452.1 | 1 | 1804 | |||
GenBank mRNA | AB037821.1 | - | 1 | 1779 | ||
AK314377.1 | - | 1 | 1154 | |||
AY013874.1 | - | 1 | 1779 | |||
BC111560.1 | - | 1 | 1142 | 40068151 | ||
KJ534913.1 | - | 1 | 953 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
