siRNA Details
Gene Symbol: PHRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286581.1 | NP_001273510.1 | 14 | 3485 | ||
NM_001286582.1 | NP_001273511.1 | 14 | 3479 | |||
NM_001286583.1 | NP_001273512.1 | 14 | 3473 | |||
NM_020901.3 | NP_065952.2 | 14 | 3482 | |||
XM_005253025.4 | XP_005253082.1 | 14 | 3443 | |||
XM_005253027.3 | XP_005253084.1 | 14 | 3431 | |||
XM_011520236.2 | XP_011518538.1 | 14 | 3479 | |||
XM_011520237.2 | XP_011518539.1 | 13 | 3326 | |||
GenBank mRNA | AB040975.1 | - | 14 | 3445 | ||
BC029651.1 | - | 1 | 1611 | |||
BC041631.1 | - | 1 | 38 | |||
BC112931.1 | - | 12 | 5215 | |||
BC136615.1 | - | 14 | 3410 | 9020605 | ||
BC144293.1 | - | 14 | 3407 | 9052813 | ||
BC144294.1 | - | 14 | 3398 | 9052814 | ||
BC144295.1 | - | 14 | 3404 | 9052815 | ||
BC146771.1 | - | 14 | 3445 | 9007218 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
