siRNA Details
Gene Symbol: WDR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017491.3 | NP_059830.1 | 4 | 605 | ||
XM_017008880.1 | XP_016864369.1 | 4 | 639 | |||
GenBank mRNA | AB010427.2 | - | 4 | 524 | ||
AB209746.1 | - | 4 | 378 | |||
AF020056.1 | - | 4 | 433 | |||
AF020260.1 | - | 4 | 433 | |||
AF274954.1 | - | 359 | ||||
AK222764.1 | - | 4 | 477 | |||
AK222897.1 | - | 4 | 509 | |||
AK225276.1 | - | 4 | 477 | |||
AK291614.1 | - | 4 | 455 | |||
AL050108.1 | - | 2 | 153 | |||
BC000201.1 | - | 4 | 404 | 3351638 | ||
BC002489.2 | - | 4 | 396 | 3051319 | ||
EU794675.1 | - | 4 | 396 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
