siRNA Details
Gene Symbol: ERCC8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000082.3 | NP_000073.1 | 12 | 1940 | ||
NM_001007233.2 | NP_001007234.1 | 13 | 2158 | |||
NM_001290285.1 | NP_001277214.1 | 11 | 1858 | |||
GenBank mRNA | AK056931.1 | - | 13 | 2131 | ||
AK226129.1 | - | 11 | 3454 | |||
U28413.1 | - | 12 | 1906 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
