siRNA Details
Gene Symbol: ZFR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_016107.3 | NP_057191.2 | 15 | 2685 | ||
GenBank mRNA | AF100742.1 | - | 5 | 680 | ||
AJ314790.1 | - | 15 | 2638 | |||
AK025481.1 | - | 3 | 378 | |||
AK295297.1 | - | 3 | 329 | |||
BC051893.1 | - | 8 | 1276 | |||
BC062986.1 | - | 8 | 1201 | |||
BC137084.1 | - | 15 | 2649 | 9021081 | ||
BC137085.1 | - | 15 | 2649 | 9021082 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
