siRNA Details
Gene Symbol: THOC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032361.3 | NP_115737.1 | 6 | 1271 | ||
XM_017009985.1 | XP_016865474.1 | 7 | 1138 | |||
GenBank mRNA | BC006849.1 | - | 6 | 1233 | 3451612 | |
BC068499.1 | - | 6 | 1190 | 5262880 |
Gene Symbol: LOC728554 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003615.2 | 6 | 1497 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
