siRNA Details
Gene Symbol: MAD2L1BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003690.1 | NP_001003690.1 | 4 | 1351 | ||
NM_014628.2 | NP_055443.1 | 3 | 1083 | |||
GenBank mRNA | AK297172.1 | - | 3 | 1104 | ||
BC002904.2 | - | 3 | 1053 | 3945348 | ||
D14811.1 | - | 3 | 1050 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
