siRNA Details
Gene Symbol: MAD2L1BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003690.1 | NP_001003690.1 | 3 | 372 | ||
NM_014628.2 | NP_055443.1 | 2 | 104 | |||
GenBank mRNA | AK297172.1 | - | 2 | 125 | ||
AL537692.3 | - | 3 | 374 | |||
BC002904.2 | - | 2 | 74 | 3945348 | ||
BE297042.1 | - | 3 | 300 | |||
BI255998.1 | - | 2 | 91 | |||
BU845738.1 | - | 3 | 410 | |||
CB991410.1 | - | 2 | 131 | |||
CR456893.1 | - | 2 | 68 | |||
D14811.1 | - | 2 | 71 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
