siRNA Details
Gene Symbol: HNRNPA2B1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002137.3 | NP_002128.1 | 9, 10 | 1188 | ||
NM_031243.2 | NP_112533.1 | 10, 11 | 1224 | |||
XM_005249729.1 | XP_005249786.1 | 10, 11 | 1206 | |||
XM_017012109.1 | XP_016867598.1 | 9, 10 | 1172 | |||
XM_017012110.1 | XP_016867599.1 | 9, 10 | 1172 | |||
GenBank mRNA | AK129839.1 | - | 9, 10 | 1142 | ||
AK289429.1 | - | 10, 11 | 1177 | |||
BC000506.2 | - | 8, 9 | 956 | 2822109 | ||
BX537494.1 | - | 8, 9 | 4021 | |||
BX641084.1 | - | 8, 9 | 4762 | |||
M29064.1 | - | 10, 11 | 1157 | |||
M29065.1 | - | 9, 10 | 1127 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
