siRNA Details
Gene Symbol: ELF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276457.1 | NP_001263386.1 | 6 | 2724 | ||
NM_001276458.1 | NP_001263387.1 | 7 | 2775 | |||
NM_001276459.1 | NP_001263388.1 | 6 | 2688 | |||
NM_006874.3 | NP_006865.1 | 7 | 2811 | |||
NM_201999.2 | NP_973728.1 | 9 | 2961 | |||
XM_005262803.2 | XP_005262860.1 | 10 | 3270 | |||
XM_005262804.2 | XP_005262861.1 | 10 | 3234 | |||
XM_005262805.2 | XP_005262862.1 | 9 | 3147 | |||
XM_006714128.2 | XP_006714191.1 | 9 | 3183 | |||
XM_006714129.3 | XP_006714192.1 | 9 | 3049 | |||
XM_011531710.2 | XP_011530012.1 | 10 | 3227 | |||
XM_011531711.2 | XP_011530013.1 | 8 | 2776 | |||
XM_011531712.2 | XP_011530014.1 | 7 | 2767 | |||
XM_011531714.2 | XP_011530016.1 | 4 | 2244 | |||
XM_017007856.1 | XP_016863345.1 | 9 | 3069 | |||
XM_017007857.1 | XP_016863346.1 | 9 | 3013 | |||
XM_017007858.1 | XP_016863347.1 | 8 | 3018 | |||
XM_017007859.1 | XP_016863348.1 | 9 | 3104 | |||
XM_017007860.1 | XP_016863349.1 | 8 | 2926 | |||
XM_017007861.1 | XP_016863350.1 | 9 | 3054 | |||
GenBank mRNA | U43188.1 | - | 9 | 2939 | ||
U43189.1 | - | 7 | 2710 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
