siRNA Details
Gene Symbol: MAPKAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006617.1 | NP_001006618.1 | 2 | 533 | ||
NM_001006618.1 | NP_001006619.1 | 2 | 533 | |||
NM_001006619.1 | NP_001006620.1 | 2 | 533 | |||
NM_024117.3 | NP_077022.1 | 2 | 533 | |||
XM_011519005.1 | XP_011517307.1 | 2 | 533 | |||
XM_011519006.2 | XP_011517308.1 | 2 | 533 | |||
XM_011519007.1 | XP_011517309.1 | 2 | 533 | |||
XM_011519008.1 | XP_011517310.1 | 2 | 533 | |||
XM_011519009.1 | XP_011517311.1 | 2 | 533 | |||
XM_017015126.1 | XP_016870615.1 | 2 | 422 | |||
XM_017015127.1 | XP_016870616.1 | 2 | 422 | |||
GenBank mRNA | AK290060.1 | - | 2 | 346 | ||
AY524429.1 | - | 2 | 530 | |||
AY524430.1 | - | 2 | 530 | |||
AY524431.1 | - | 2 | 530 | |||
AY524432.1 | - | 2 | 530 | |||
AY633624.1 | - | 2 | 500 | |||
AY633625.1 | - | 2 | 507 | |||
AY633626.1 | - | 2 | 507 | |||
BC002326.2 | - | 2 | 502 | 2823015 | ||
BC003044.1 | - | 2 | 502 | 2823015 | ||
BX438260.2 | - | 2 | 532 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
