siRNA Details
Gene Symbol: TMEM132B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286219.1 | NP_001273148.1 | 5 | 1110 | ||
NM_052907.3 | NP_443139.2 | 9 | 2363 | |||
XM_011537852.2 | XP_011536154.1 | 9 | 2420 | |||
XM_011537854.2 | XP_011536156.1 | 5 | 7093 | |||
XM_017018767.1 | XP_016874256.1 | 10 | 2529 | |||
GenBank mRNA | AB067493.1 | - | 4 | 900 | ||
AK093097.1 | - | 5 | 1066 | |||
BC113366.1 | - | 5 | 987 | 8322418 | ||
BC113368.1 | - | 5 | 987 | 8322420 | ||
BC131621.1 | - | 5 | 1027 | 40117411 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
