siRNA Details
Gene Symbol: RFX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206691.1 | NP_001193620.1 | 12 | 1324 | ||
NM_032491.5 | NP_115880.2 | 8 | 990 | |||
NM_213594.2 | NP_998759.1 | 12 | 1571 | |||
GenBank mRNA | AB044245.1 | - | 12 | 1289 | ||
AB095365.1 | - | 12 | 1319 | |||
AB095366.1 | - | 12 | 1571 | |||
AB195784.1 | - | 2 | 164 | |||
AB195785.1 | - | 2 | 86 | |||
AF332192.1 | - | 8 | 984 | |||
AK074913.1 | - | 9 | 902 | |||
AK291445.1 | - | 12 | 1556 | |||
AK315698.1 | - | 8 | 970 | |||
AL833921.1 | - | 5 | 394 | |||
AY102009.1 | - | 12 | 1224 | |||
BC028582.2 | - | 8 | 993 | 4836964 | ||
BC030644.1 | - | 8 | 965 | 4825079 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
