siRNA Details
Gene Symbol: RFX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001206691.1 | NP_001193620.1 | 6 | 648 | ||
NM_032491.5 | NP_115880.2 | 2 | 314 | |||
NM_213594.2 | NP_998759.1 | 6 | 895 | |||
GenBank mRNA | AB044245.1 | - | 6 | 613 | ||
AB095365.1 | - | 6 | 643 | |||
AB095366.1 | - | 6 | 895 | |||
AF332192.1 | - | 2 | 308 | |||
AK074913.1 | - | 3 | 226 | |||
AK291445.1 | - | 6 | 880 | |||
AK302837.1 | - | 6 | 619 | |||
AK315698.1 | - | 2 | 294 | |||
AY102009.1 | - | 6 | 548 | |||
BC028582.2 | - | 2 | 317 | 4836964 | ||
BC030644.1 | - | 2 | 289 | 4825079 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
