siRNA Details
Gene Symbol: RTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021136.2 | NP_066959.1 | 8, 9 | 2504 | ||
NM_206852.2 | NP_996734.1 | 6, 7 | 899 | |||
XM_017021578.1 | XP_016877067.1 | 6, 7 | 788 | |||
GenBank mRNA | AK128113.1 | - | 3, 4 | 2364 | ||
AK290534.1 | - | 6, 7 | 899 | |||
AK297492.1 | - | 7, 8 | 806 | |||
BC000314.2 | - | 6, 7 | 684 | |||
BC003003.1 | - | 6, 7 | 684 | |||
BC090862.1 | - | 8, 9 | 2419 | 6179803 | ||
BC111694.1 | - | 9, 10 | 2194 | 40035553 | ||
BX449967.2 | - | 6, 7 | 887 | |||
CR457030.1 | - | 6, 7 | 591 | |||
L10333.1 | - | 8, 9 | 2417 | |||
L10334.1 | - | 6, 7 | 1325 | |||
L10335.1 | - | 6, 7 | 619 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
