siRNA Details
Gene Symbol: RTN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021136.2 | NP_066959.1 | 9 | 2584 | ||
NM_206852.2 | NP_996734.1 | 7 | 979 | |||
XM_017021578.1 | XP_016877067.1 | 7 | 868 | |||
GenBank mRNA | AK128113.1 | - | 4 | 2444 | ||
AK290534.1 | - | 7 | 979 | |||
AK297492.1 | - | 8 | 886 | |||
BC000314.2 | - | 7 | 764 | |||
BC003003.1 | - | 7 | 764 | |||
BC090862.1 | - | 9 | 2499 | 6179803 | ||
BC111694.1 | - | 10 | 2274 | 40035553 | ||
L10333.1 | - | 9 | 2497 | |||
L10334.1 | - | 7 | 1405 | |||
L10335.1 | - | 7 | 699 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
