siRNA Details
Gene Symbol: WDR89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008726.2 | NP_001008726.1 | 4 | 1207 | ||
NM_001258272.1 | NP_001245201.1 | 3 | 1603 | |||
NM_080666.3 | NP_542397.1 | 3 | 1098 | |||
XM_011536383.2 | XP_011534685.1 | 2 | 1137 | |||
XM_011536385.2 | XP_011534687.1 | 4 | 1105 | |||
GenBank mRNA | AF115513.1 | - | 3 | 1030 | ||
AK095324.1 | - | 3 | 1046 | |||
BC010698.1 | - | 2 | 990 | 3870073 | ||
BX248008.1 | - | 3 | 1087 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
