siRNA Details
Gene Symbol: CDR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001802.1 | NP_001793.1 | 3 | 529 | ||
GenBank mRNA | AK292273.1 | - | 3 | 276 | ||
BC017503.1 | - | 3 | 511 | 4873337 | ||
BG750955.1 | - | 3 | 512 | |||
D12981.1 | - | 3 | 205 | |||
M63256.1 | - | 3 | 389 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
