siRNA Details
Gene Symbol: SS18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007559.2 | NP_001007560.1 | 3 | 264 | ||
NM_001308201.1 | NP_001295130.1 | 3 | 239 | |||
NM_005637.3 | NP_005628.2 | 3 | 264 | |||
XM_006722527.2 | XP_006722590.1 | 3 | 226 | |||
XM_011526145.1 | XP_011524447.1 | 3 | 232 | |||
XM_011526147.2 | XP_011524449.1 | 5 | 515 | |||
XM_011526148.2 | XP_011524450.1 | 5 | 502 | |||
XM_011526149.2 | XP_011524451.1 | 5 | 527 | |||
XM_011526150.2 | XP_011524452.1 | 4 | 387 | |||
XM_011526151.2 | XP_011524453.1 | 5 | 516 | |||
XM_011526152.2 | XP_011524454.1 | 4 | 387 | |||
GenBank mRNA | AF244972.1 | - | 3 | 201 | ||
AF343880.1 | - | 3 | 198 | |||
AK289510.1 | - | 3 | 242 | |||
AK296949.1 | - | 3 | 222 | |||
AK301504.1 | - | 3 | 269 | |||
AK304305.1 | - | 4 | 380 | |||
AK307923.1 | - | 2 | 155 | |||
AK316302.1 | - | 3 | 214 | |||
AK316338.1 | - | 4 | 333 | |||
AU279615.1 | - | 3 | 269 | |||
BC096221.1 | - | 3 | 201 | 40004908 | ||
BC096222.1 | - | 3 | 201 | 40004909 | ||
BC096223.1 | - | 3 | 201 | 40004911 | ||
BC096224.1 | - | 3 | 201 | 40004912 | ||
CR542088.1 | - | 3 | 198 | |||
CR542103.1 | - | 3 | 198 | |||
DC378335.1 | - | 3 | 239 | |||
X79201.1 | - | 3 | 201 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
