siRNA Details
Gene Symbol: TTYH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032646.5 | NP_116035.5 | 8 | 920 | ||
NM_052869.1 | NP_443101.1 | 2 | 730 | |||
XM_005257824.3 | XP_005257881.1 | 8 | 848 | |||
GenBank mRNA | AF319952.1 | - | 8 | 899 | ||
AK091085.1 | - | 6 | 601 | |||
AK126955.1 | - | 2 | 1138 | |||
AK296511.1 | - | 8 | 843 | |||
AK315163.1 | - | 8 | 906 | |||
BC005168.1 | - | 8 | 895 | |||
BC107492.1 | - | 8 | 916 | 40033786 | ||
BC107602.1 | - | 8 | 916 | 40033791 | ||
BX647912.1 | - | 2 | 730 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
