siRNA Details
Gene Symbol: TTYH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032646.5 | NP_116035.5 | 14 | 3232 | ||
NM_052869.1 | NP_443101.1 | 8 | 3042 | |||
XM_005257824.3 | XP_005257881.1 | 14 | 3160 | |||
GenBank mRNA | AF319952.1 | - | 14 | 3209 | ||
AK058197.1 | - | 3 | 1932 | |||
AK091085.1 | - | 12 | 2913 | |||
AK126955.1 | - | 8 | 3450 | |||
AL831858.1 | - | 4 | 1978 | |||
BC004233.2 | - | 1 | 1310 | |||
BX647912.1 | - | 8 | 3042 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
