siRNA Details
Gene Symbol: ZNF473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_017026574.1 | XP_016882063.1 | 3 | 4307 | ||
GenBank mRNA | AB032967.1 | - | 5 | 3064 | ||
AF454744.1 | - | 4 | 3019 | |||
AL080143.1 | - | 1 | 1372 | |||
BX648093.1 | - | 5 | 3108 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
