siRNA Details
Gene Symbol: RPL13A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270491.1 | NP_001257420.1 | 7 | 720 | ||
NM_012423.3 | NP_036555.1 | 8 | 796 | |||
NR_073024.1 | 8 | 786 | ||||
GenBank mRNA | AB082924.1 | - | 5 | 575 | ||
AK056837.1 | - | 3 | 1933 | |||
AK130605.1 | - | 8 | 743 | |||
AK291120.1 | - | 8 | 742 | |||
AK297859.1 | - | 6 | 546 | |||
BC000514.2 | - | 8 | 725 | 2822802 | ||
BC004900.2 | - | 7 | 716 | |||
BC032107.2 | - | 8 | 731 | |||
BC062537.1 | - | 7 | 722 | 4908022 | ||
BC070223.1 | - | 8 | 742 | 6736442 | ||
BC071929.1 | - | 7 | 726 | 5808487 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
