siRNA Details
Gene Symbol: PRDM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022114.3 | NP_071397.3 | 14 | 3280 | ||
NM_199454.2 | NP_955533.2 | 14 | 3280 | |||
XM_005244772.4 | XP_005244829.1 | 14 | 3432 | |||
XM_005244773.4 | XP_005244830.1 | 14 | 3436 | |||
XM_005244774.4 | XP_005244831.1 | 14 | 3435 | |||
XM_006710814.3 | XP_006710877.1 | 14 | 3430 | |||
XM_011541945.2 | XP_011540247.1 | 13 | 3053 | |||
XM_017002050.1 | XP_016857539.1 | 14 | 3280 | |||
GenBank mRNA | AB051462.2 | - | 14 | 3230 | ||
AB078876.1 | - | 14 | 3242 | |||
AF294278.1 | - | 14 | 3261 | |||
AK309078.1 | - | 13 | 2976 | |||
BC161614.1 | - | 14 | 3230 | 8860117 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
