siRNA Details
Gene Symbol: PRDM16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022114.3 | NP_071397.3 | 17 | 8404 | ||
NM_199454.2 | NP_955533.2 | 17 | 8347 | |||
GenBank mRNA | AB051462.2 | - | 17 | 8354 | ||
AK093853.1 | - | 1 | 1740 | |||
AK094178.1 | - | 1 | 2183 | |||
BC161614.1 | - | 17 | 8354 | 8860117 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
