siRNA Details
Gene Symbol: ADAMTSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288607.1 | NP_001275536.1 | 13 | 2181 | ||
NM_001288608.1 | NP_001275537.1 | 13 | 2367 | |||
NM_019032.5 | NP_061905.2 | 12 | 2298 | |||
NM_025008.4 | NP_079284.2 | 12 | 2298 | |||
XM_011509644.2 | XP_011507946.1 | 12 | 2329 | |||
XM_011509645.2 | XP_011507947.1 | 11 | 2257 | |||
XM_011509648.2 | XP_011507950.1 | 12 | 2283 | |||
XM_011509649.2 | XP_011507951.1 | 12 | 2337 | |||
XM_011509650.2 | XP_011507952.1 | 12 | 2337 | |||
XM_011509651.2 | XP_011507953.1 | 6 | 1079 | |||
XM_011509652.1 | XP_011507954.1 | 6 | 785 | |||
XM_017001506.1 | XP_016856995.1 | 13 | 2738 | |||
XM_017001507.1 | XP_016856996.1 | 4 | 520 | |||
GenBank mRNA | AF217974.1 | - | 1 | 100 | ||
AF370411.1 | - | 6 | 834 | |||
AK092509.1 | - | 3 | 418 | |||
AK293037.1 | - | 2 | 230 | |||
AY358122.1 | - | 11 | 2112 | |||
BC027478.1 | - | 5 | 737 | |||
BC071852.1 | - | 4 | 594 | |||
BC094811.1 | - | 4 | 594 | |||
BC140800.1 | - | 12 | 2219 | 9021668 | ||
BC144581.1 | - | 12 | 2033 | 9053111 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
