siRNA Details
Gene Symbol: LYST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000081.3 | NP_000072.2 | 5 | 808 | ||
NM_001301365.1 | NP_001288294.1 | 5 | 1245 | |||
XM_011544031.1 | XP_011542333.1 | 5 | 808 | |||
XM_011544032.1 | XP_011542334.1 | 5 | 1211 | |||
XM_011544033.2 | XP_011542335.1 | 5 | 958 | |||
XM_011544034.1 | XP_011542336.1 | 5 | 808 | |||
XM_011544035.2 | XP_011542337.1 | 5 | 808 | |||
XM_011544037.2 | XP_011542339.1 | 5 | 808 | |||
XM_011544039.2 | XP_011542341.1 | 5 | 808 | |||
XM_011544040.2 | XP_011542342.1 | 5 | 808 | |||
XM_017000150.1 | XP_016855639.1 | 5 | 808 | |||
XM_017000151.1 | XP_016855640.1 | 5 | 808 | |||
GenBank mRNA | AK307457.1 | - | 5 | 1211 | ||
BC144702.1 | - | 5 | 1109 | |||
U67615.1 | - | 5 | 815 | |||
U70064.1 | - | 5 | 1178 | |||
U72192.1 | - | 5 | 1178 | |||
U84744.1 | - | 5 | 1178 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
