siRNA Details
Gene Symbol: CALM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001305624.1 | NP_001292553.1 | 7 | 801 | ||
NM_001305625.1 | NP_001292554.1 | 6 | 660 | |||
NM_001305626.1 | NP_001292555.1 | 5 | 4015 | |||
NM_001743.5 | NP_001734.1 | 6 | 755 | |||
GenBank mRNA | AK055130.1 | - | 5 | 2472 | ||
BC003354.1 | - | 6 | 631 | 2899899 | ||
BC006464.1 | - | 6 | 633 | 3542044 | ||
BC008437.1 | - | 6 | 649 | 4092978 | ||
BC017385.1 | - | 6 | 647 | 3681165 | ||
BC018677.1 | - | 6 | 648 | 4656551 | ||
BC026065.1 | - | 6 | 647 | 3683323 | ||
D45887.1 | - | 6 | 643 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
