siRNA Details
Gene Symbol: VPS54 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005739.1 | NP_001005739.1 | 22 | 2893 | ||
NM_016516.2 | NP_057600.2 | 22 | 2929 | |||
XM_006712029.3 | XP_006712092.1 | 21 | 2789 | |||
XM_017004316.1 | XP_016859805.1 | 21 | 2609 | |||
GenBank mRNA | AF102177.1 | - | 21 | 2786 | ||
AK002205.1 | - | 10 | 1034 | |||
AK025545.1 | - | 16 | 1821 | |||
AK094019.1 | - | 7 | 1340 | |||
AK124436.1 | - | 11 | 4077 | |||
AK292669.1 | - | 23 | 2937 | |||
AK292889.1 | - | 23 | 2988 | |||
AL137604.1 | - | 1 | 48 | |||
AL359939.1 | - | 9 | 751 | |||
AY444798.1 | - | 21 | 2786 | |||
BC030275.1 | - | 22 | 2896 | 3917367 | ||
BC041868.1 | - | 19 | 2897 | 5271464 | ||
CR749701.1 | - | 9 | 751 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
